Learn More
Oxyrase Cryobroth Preservation Product, 10Ml Bottle 1/Pk
Supplier: Oxyrase PRCRYO
Store and preserve microorganisms under cryogenic conditions. Optimizes cell preservation and recovery at ultra-low temperatures. Quantitative recovery. Designed to be liquid at room temperature. Easy to handle, aliquot and inoculate.
This item is not returnable.
View return policy