Learn More
ProteinSimple 66-440 kDa Separation Module, 8 x 25 capillary cartridges
Supplier: ProteinSimple SMW008
Compatible with Jess, Abby and Wes systems. Includes separation box with Capillary Cartridges, 66-440 kDa Pre-filled Plates (pre-filled with Separation Matrix 2, Stacking Matrix 2, Split Running Buffer 2 and Matrix Removal Buffer), Sample Buffer, Wash Buffer, plus Standard Pack 1 clamshell with DTT, FL Standard master mix with 29 kDa System Control protein. Compatible with Detection module - sold separately. Sizing: 8x25 capillaries per cartridge (200 data points).
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.