Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Tocris Bioscience™ CRID3 sodium salt
Potent NLRP3 inflammasome inhibitor; inhibits IL-1β production
$294.64 - $1229.60
Specifications
Purity | 98% |
---|---|
Description | CRID3 sodium salt |
Molecular Weight (g/mol) | 426.46 |
Product Type | CRID3 sodium salt |
Description
Potent NLRP3 inflammasome inhibitor; inhibits IL-1β (IC50 = 7.2 nM), IL-1α (IC50 = 12-18 nM) and IL-18 (IC50 = 10.3 nM) production. Selective for NLRP3 inflammasome over NLRC4 inflammasome and Toll-like receptor signaling. Inhibits inflammasome activation by soluble and crystalline NLRP3 stimuli. Reduces severity of experimental autoimmune encephalomyelitis, skin inflammation and airway inflammation in mice. Also glutathione S-transferase omega 1 inhibitor. Orally bioavailable.Specifications
98% | |
CRID3 sodium salt | |
426.46 | |
CRID3 sodium salt |
For Research Use Only
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title