Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Ansell™ CPP Vinyl Apron 56-100

Heavy-duty protection.
Supplier: Ansell™ 56 100 50950101/EMD
This item is not returnable.
View return policy
Description
- Heavy-duty PVC construction
- Three piece ties
- Resistance to chemicals, fats, oils, greases, punctures and abrasions
Specifications
Apron | |
Vinyl | |
3 piece ties | |
One piece | |
Food processing and preparations, Chemical processing, oil refining, and petrochemicals, janitorial and maintenance work | |
50 in. |
Universal | |
Green | |
Bib | |
20 mil | |
50 x 35 in. | |
35 in. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title