Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Corning™ Flexible Packaging Systems: MPC Connector Tubing
Clear tubing; female MPC connector
Supplier: Corning™ 9170012
Description
- Chemically resistant
- Heat sealable
- Flexible
Specifications
Connector tubing | |
cGMPs, FDA Regulation 21 CFR part 820 and ISO 13485 Quality | |
Pinch clamp and filling bell, 0.25 in. barb |
Fits tubing 60.96 cm | |
Clear |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction