Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Corning™ Flexible Packaging Systems: HDPE Media Bag Tote
Meet FDA requirements as 510K Medical Device
Supplier: Corning™ 99CARRIER
Description
Nestable/stackable container constructed of rigid, high-density polyethylene (HDPE)
- With C-Flex tubing and two Luer-Lok fittings
- Ideal for cleanroom use and as storage containers
- Accommodates up to 25L of media or process reagents
- Maximum weight capacity: 45 lb. (20.41 kg)
Biopharmaceutical
Specifications
Media Bag Tote | |
HDPE |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction