Learn More
Cole-Parmer Stopper Hole Boring Set, Chrome-Plated Steel; 12 Borers/Set
Supplier:  Cole-Parmer 0629890
Encompass_Preferred
This chrome-plated steel cork borer set stays sharp. Flat blades permit easy boring of lightweight plastic, cork stoppers, and spongy articles. Set consists of twelve borers with outside diameters of 4, 6, 7.5, 8.5, 10, 12, 13, 15, 17, 18.5, 19, and 20.5 mm.
Catalog No. NC9546789