Learn More
USA BLUE BOOK CorePRO Jr. 15' Sampler, Complete Set
Supplier: USA BLUE BOOK 41340
CorePRO Samplers provide more strength and a better sample! We got tired of hearing complaints about the flimsiness of other core samplers. CorePRO samplers provide much more strength and a better sample. This proprietary design uses three 5-ft long sections that screw together for portability. The bottom section has a built-in foot valve to hold the sample for blanket observation. Tubes have tape markings every foot without numbers (order optional self-stick waterproof numbers below). You can also purchase individual midsections for lengthening beyond 15-ft or for repair if you run over it with a truck. The famous 15-ft CorePRO Jr. is relatively flexible with a 1"OD. It has improved joints with stronger fittings and no rivets. Connect up to four sections for 20-ft maximum length. Samplers offer economy plus durability.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.