Learn More
USA BLUE BOOK COREPRO 15' SAMPLER/3 SECT
Supplier: USA BLUE BOOK 71475
15 FT COREPRO SAMPLER (3 SECTIONS),Improves on the Sludge Judge and Coretaker,made of diameter LEXAN,Threaded tube connections are made of polypropylene and PVC,can connect up to 5 sections for 25-ft total maximum length.