Learn More
Science First™ Copper Plating Set
Demonstrates technique used for copper plating.
Supplier: Science First™ 6650215
Includes: Glass battery jar, copper anode, brass connecting posts (2), wire connectors with alligator clips (2), copper sulfate (500g), MSDS, and paper clips
Specifications
Copper Plating Set | |
Shows copper plating technique | |
Glass battery jar, copper anode, 2 Brass connecting posts, 2 Wire connectors with alligator clips, 500g Copper sulfate, MSDS and paper clips |