Learn More
Copan Diagnostics Copan Diagnostics Eswab™ Patented Sample Collection and Delivery System + Rayon Swab
SDP
Liquid based collection and transport device for aerobic, anaerobic, and fastidious bacteria for up to 48 hours. Polypropylene screw-cap tube filled with 1mL of Liquid Amies medium, plus one white regular size Nylon flocked swab and one pink regular size rayon swab.
Supplier:  Copan Diagnostics 486C
Catalog No. 23-600-906
Edge
Add to Cart
This item is not returnable. View return policy
Description
Description
  • Standard ESwab™ product with additional rayon swab Maintains viability of aerobic, anaerobic, and fastidious bacteria for up to 48 hours
  • Using the patented FLOQSwab technology ESwab elutes the patient sample upon contact with the medium
  • Better growth and pathogen recovery - elutes entire sample upon contact with transport media
  • Equivalent sample distribution across all media plates
  • Better gram slides - homogenous preparation without specimen debris
  • Simplify and streamline - consolidate less optimal collection swabs into one superior system
  • One swab for up to 10 tests
  • Allows for easy automated specimen processing
Specifications
Specifications
Standard ESwab product with additional rayon swab