Learn More
CoorsTek™ Porous Cups
SCI_ED
Supplier:  CoorsTek™ 60495/EMD
 View more versions of this product
Catalog No. S43347
Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.
This item is not returnable. View return policy
Description
Description
These unglazed porcelain cylinders are designed for use with voltaic cells. Available in two popular sizes.
Specifications
Specifications
Voltaic Cell
For mechanical and earth science uses include filtration, diffusion, pressure studies, and underground liquid containment studies
900°C to 1400°C
3.9 in.
2.04 in.
Porcelain, Ceramic
1652°F to 284°F
Porous Cup
10 cm
5.2 cm
Product Suggestions
Product Suggestions