Learn More
ORS Nasco Cooper™ Ball Pein Hammers
For striking chisels and punches, and for riveting, shaping, and straightening unhardened metal
Supplier:  ORS Nasco 18411520
Catalog No. 19-313-581
Item Discontinued This item has been discontinued and is no longer available. Please reach out to your sales representative.
Description
Description
  • Ground and polished, black head
  • Permabond™ construction between head and handle
Specifications
Specifications
Ball Peen Hammer
15 in.
Steel
1.5 lb.
0.68 kg
Hickory
38 cm
1.75 lb.
0.79 kg