Learn More
Andwin Scientific CONVERTORS DISP TOWELS
Supplier: Andwin Scientific CV3520
The Absorbent Drape Towel are constructed of polylined material that generates an impenetrable bottom. This product is sterile. The towel can be used for general laboratory cleanup or protection of cleanroom surfaces and equipments. It measures 45.7 x 66 cm (18 x 26").
This item is not returnable.
View return policy