Learn More
Mopec™ Control Slides, IHC SMI-31
SDP
All control slides are on Mopec™ IHC Enhanced Hydrophilic Adhesive Slides using Formalin Fixed Paraffin Embedded (FFPE) human tissue.
Supplier:  Mopec™ SC520100
 View more versions of this product
Catalog No. 22-452-42
Explore available promotions
Description
Description
  • Product specific reactivity is guaranteed for one year from date of receipt when stored properly at 60°F to 80°F in a humidity controlled and light-deprived environment
  • Case of 25 contains 24 unstained slides and 1 stained slide
  • Case of 100 contains 98 unstained slides and 2 stained slides
Specifications
Specifications
Uncoated
98 Unstained, 2 Stained
Human
IHC Control Slide
No
1 mm
Glass
Normal
75 x 25 mm
Product Suggestions
Product Suggestions