Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Congo Red, Certified, Sigma-Aldrich™

Catalog Number SDC6277
Encompass
Click to view available options
Quantity:
25 g
573-58-0
C32H22N6Na2O6S2
696.664
IQFVPQOLBLOTPF-UHFFFAOYSA-L
Cosmos Red
11313
CHEBI:34653
disodium;4-amino-3-[[4-[4-[(1-amino-4-sulfonatonaphthalen-2-yl)diazenyl]phenyl]phenyl]diazenyl]naphthalene-1-sulfonate
C1=CC=C2C(=C1)C(=CC(=C2N)N=NC3=CC=C(C=C3)C4=CC=C(C=C4)N=NC5=C(C6=CC=CC=C6C(=C5)S(=O)(=O)[O-])N)S(=O)(=O)[O-].[Na+].[Na+]
573-58-0
C32H22N6Na2O6S2
696.664
IQFVPQOLBLOTPF-UHFFFAOYSA-L
Cosmos Red
11313
CHEBI:34653
disodium;4-amino-3-[[4-[4-[(1-amino-4-sulfonatonaphthalen-2-yl)diazenyl]phenyl]phenyl]diazenyl]naphthalene-1-sulfonate
C1=CC=C2C(=C1)C(=CC(=C2N)N=NC3=CC=C(C=C3)C4=CC=C(C=C4)N=NC5=C(C6=CC=CC=C6C(=C5)S(=O)(=O)[O-])N)S(=O)(=O)[O-].[Na+].[Na+]

Certified by the Biological Stain Commission. Certified for use as a counterstain for alum based hematoxylin and in the Highman or the Churukian modification of Puchtler's amyloid stain

Grade Certified
Quantity 25 g
Chemical Name or Material Congo Red
Provide Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit