Learn More
Market Lab MarketLab Rotating Vial Rack, 10"Dia x 2"H, 1 each
Supplier: Market Lab 3159
Large capacity, small footprint saves valuable refrigerator space. Color-coded segments ensure correct lot and choice of calibrators and controls by instrument, reducing time and preventing lot number mix-ups. Segments sit securely on a rotating base. MarketLab exclusive.