Learn More
Ace Glass, Inc. 10ML SPEC GRAV BTL W/STOPPER
Supplier: Ace Glass, Inc. 542018
Guy-Lussac type 10mL specific gravity bottle with 7/18 stopper, unadjusted for calibration in the laboratory. When adjusted, the bottle is suitable for ASTM D 369.
This item is not returnable.
View return policy