Learn More
Sigma Aldrich Fine Chemicals Biosciences Freund’s Adjuvant, Complete, MFCD00131105, 10 mL
Supplier: Sigma Aldrich Fine Chemicals Biosciences F588110ML
Synonym: FCA. Cell suspension. Each mL contains 1 mg of Mycobacterium tuberculosis (H37Ra, ATCC 25177), heat killed and dried, 0.85 mL paraffin oil and 0.15 mL mannide monooleate. Antigens (preferably in saline) are typically mixed with an equal volume of the adjuvant to form an emulsion. Freund′s Adjuvants may be used to produce water-in-oil emulsions of immunogens. Antigens in water-in-oil emulsions stimulate high and long-lasting antibody responses which can be attributed to the slow release of antigen.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.