Learn More
Grainger Barrel Coliwasa Waste Sampler 460Ml
Supplier:  Grainger 9EX94
Encompass_Preferred
Barrel Coliwasa, 460mL, 39.5 in. L, Sell Pack: 1 EA, It has a 2 in. stick-out on bottom for ready-fill sampling of viscous liquids and semi-liquids, Made of translucent, unmodified polypropylene for superior chemical resistance
Catalog No. NC9754389