Learn More
MilliporeSigma™ Supelco™ Column for Flash-Chromatography Assembly with Threaded Joints and Fritted Disc (Porosity C), Joint Size: ST/NS 45/40
Flash chromatography is useful for rapid, preparative separations with moderate resolutions.
Supplier:  MilliporeSigma™ Supelco™ Z4165411EA
 View more versions of this product
Catalog No. 11-100-8834
Add to Cart
Description
Description
Specifications
Specifications
508 mm
Flash Chromatography
1000 mL
57 mm
Flash Chromatography
ST/NS 45/40