Learn More
MilliporeSigma™ Supelco™ Column for Flash-Chromatography Assembly with Ball Joints, Standard, Joint Size: SJ 65/40
Flash chromatography is useful for rapid, preparative separations with moderate resolutions.
Supplier:  MilliporeSigma™ Supelco™ Z1070181EA
Catalog No. 11-100-8868
Add to Cart
Description
Description
Specifications
Specifications
572 mm
Flash Chromatography
600 mL
44 mm
Flash Chromatography
SJ 65/40