Learn More
Helena Laboratories™ ColoScreen™ FOB Test Kit
Convenient triple slide provides three successive tests in one unit
Specifications
Clia Complexity | Waived |
---|---|
For Use With (Application) | Fecal Occult Blood Testing |
Quantity | 100 Pack |
Catalog Number | Mfr. No. | Quantity | Includes | Type | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Includes | Type | Price | Quantity & Availability | |||
23-038-046
|
Helena Laboratories™
5071 |
100 Pack | 100 Triple unit ColoSreen slides with on-slide Colocheck monitors, 6 x 15mL ColoScreen developer, 300 specimen applicator, 100 Coloscreen mailing envelopes, 100 patient instruction sheets | ColoScreen Office Pack |
Pack of 100 for $385.00
|
N/A | |||
Due to product restrictions, please Sign In to purchase or view availability for this product. | |||||||||
23-038-049
|
Helena Laboratories™
5082 |
100 Pack | 34 Triple unit ColoScreen slides with on-slide, ColoCheck monitors, 2 x 15mL ColoScreen developer, 100 specimen applicator | ColoScreen Lab Pack |
Pack of 100 for $175.00
|
N/A | |||
Due to product restrictions, please Sign In to purchase or view availability for this product. | |||||||||
Description
- CLIA Complexity: Waived
- Ideal for physician's office or clinic use
- Classic guaiac slide format
- Office Pack allows slides, sample applicators and mailing envelopes to be conveniently distributed to patients, who can then prepare test slides in the privacy of their homes
- Clinic Pack is designed for labs preferring the triple unit slides
- Lab Pack is provided in order that each laboratory can stock the volume that best fits its demands
- Each single slide in the triple unit has two test areas so that multiple sites from each specimen may be tested, increasing accuracy
Specifications
Waived | |
Fecal Occult Blood Testing | |
100 Pack |