Learn More
Thermo Scientific Chemicals ColorpHast™ pH Strips, Intermediate, three squares with different indicators on each strip
Supplier:  Thermo Scientific Chemicals 035234LQ
 View more versions of this product
Catalog No. AA35234LQ
Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.
Description
Description
ColorpHast™ pH Strips, Universal, four squares with different indicators on each strip, 0-14 pH range is used in analytical laboratory to measure the pH readings in seconds in drinking water, saliva and in chemical reactions. It is utilized for quick and accurate measurement and saves our time and trouble.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
5 to 10
1 Unit
Intermediate pH Strip
Product Suggestions
Product Suggestions