Learn More
Vernier™ Color Mixer Kit
Experiments in additive and subtractive color mixing can be easily and conveniently carried out using the Vernier Color Mixer Kit.
Supplier: Vernier™ CMOEK
Includes: Color Mixer Kit
Description
- The Color Mixer Kit consists of a three-color LED illuminator with power supply, a lens, and a double-sided screen. The intensity of the red, blue and green LEDs can be smoothly controlled from the light source.
- Requires a track to serve as an optics bench.
Warranty and Services
5-year limited warranty
Specifications
Kit | |
Educational use only | |
Three color LED illuminator |