Learn More
Color-Coded Spring Scales
Clear plastic tube allows students to see the action of the spring.
Supplier: Si Metric Manufacturing Limited 39630
Description
Series of eight color-coded tubular spring scales can be used for both force and mass measurements.
- Calibrated in both Newtons and grams with a beveled top for use on an inclined plane
- Incorporates a unique zero adjustment
Order Info
Available as a set of six colors (Cat. No. S02321A).
Specifications
White | |
4+ | |
6.6 lb., 3 kg | |
Spring Scale |