Learn More
Uline Cold Packs 8oz, 6/case
Supplier: Uline S7361
Reusable cold packs with non-toxic gel for temporary cooling during transport, gel remains pliable when frozen to mold around contents in durable plastic casing.