Learn More
Chemglass Life Sciences Midget Impinger, Graduated, Complete, 24/40 Joint Size, 12/5 Tip
Supplier: Chemglass Life Sciences CG182002
Midget Impinger, Graduated, Complete, 24/40 Joint Size, 12/5 Tip. Micro impinger designed for sampling of small volumes of air at low jet velocity, as per EPA Method 6. Impinger bottle is graduated from 5 to 30ml in 5ml subdivisions. Both the bottle and stopper have 24/40 standard taper joints. The orifice at the impinger tip is 1mm, passing 0.09 to 0.11 CFM of air at 12in H20 vacuum. Equipped with 12/5 ball joints.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.