Learn More
Chemglass Life Sciences Extraction Apparatus, Jumbo, Size B
Supplier: Chemglass Life Sciences CG137302
Extraction Apparatus, Jumbo, Size B. Jumbo Soxhlet extraction apparatus equipped with a Duran flange at the top of the extractor. Flange is formed with a groove in the flat portion of the mating surface to accept a PTFE covered silicone o-ring to permit grease free assembly. The adapter between the condenser and extractor has a top outer joint and a Duran flange which attaches to the extractor body with a CG-141 stainless steel clamp. Complete apparatus consists of a condenser, extractor, round bottom flask, adapter, PTFE covered o-ring and clamp. PTFE sleeves and heating mantles must be ordered separately.Extractor Capacity (mL) 1,500 Extractor I.D. (mm) 95 Extractor Flange Size 100 Extractor Lower Joint 45/50 Condenser Length (mm) 400 Paper Thimble Size (mm) 81 x 230 Flask Capacity (mL) 3,000
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.