Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Haz-Mat Decontaminable SKED™Stretchers
Can be stored flat on pallets with straps allowing quick movement of large number of stretchers
Supplier: Cmc Rescue 752507
Description
- Pallets of 50 to 200 HMD stretchers can be moved by trucks or airlifted by helicopters, thus facilitating rapid movement of patients
- Allow patient or downed rescuer to be quickly rolled onto the stretcher and pulled out of the hot zone by rescuers who are rope length away
- Chemical resistant polypropylene straps
- 2 in. side-release buckles are easy to operate even with thick gloves
- Helpful in mass casualty or hazardous material incidents
Specifications
Stretcher System | |
Mass casualty and hazardous material incidents | |
3 lb. |
Yellow | |
Polypropylene straps | |
6.1 kg |
Provide Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction