Learn More
STEMCELL Technologies ClonaCell™-CHO ACF Medium, Size: 90 mL
Supplier: STEMCELL Technologies 03816
Serum-free, animal component-free semi-solid medium with supplements for selection and cloning of transfected CHO cells