Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CleanSpace™ CleanSpace2™ Hi Capacity Power Unit

Battery-powered, positive pressure air-purifying respirator designed to provide comfortable respiratory protection in the workplace.

Supplier:  CleanSpace™ PAF1034

Catalog No. 17-800-600

This item is not returnable. View return policy

Description

Description

  • Contains: harness, neck supports, battery charger, flow test cap, mask fit cap, carry bag
  • Light weight and comfortable
  • AirSensit™ System for mask pressure control and breath responsive airflow
  • One-button system and standby and auto-start/stop mode for quick don/doffing
  • Easy to use support harness
  • Altitude compensated filter blocked detection
  • Long operating time: up to 8 hours and quick battery recharging:< 2hours
  • Can be tested by Quantitatively Mask Fit (PortaCount) Tests
  • Compatible with all CleanSpace™ Half Masks
  • Compatible with all CleanSpace™ filters: Particulate
Specifications

Specifications

8 hours
220 L/min.
500 g
PAPR Assembly, No Headtop
PAF-1034
Product Suggestions

Product Suggestions

SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit