Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Fisherbrand™ Cleanroom Sock Recycling Solution
By participating in the Fisherbrand Cleanroom Sock Recycling Solution program, your cleanroom socks will be diverted from the landfill. The box is made from recycled materials and includes a prepaid shipping label. Fill it at your own rate and ship it back for recycling when full.
Description
- Recycling solution for disposable cleanroom socks
- All brands of disposable cleanroom socks are accepted
- Eco-friendly Boxes: Made from recycled materials and easy to assemble
- Collection boxes include a self-locking mechanisms and prepaid shipping label
- Do not recycle biological materials, infectious waste, bloodborne pathogens, hazardous materials or socks exposed to more than 10% chemical exposure of the surface area
- Available in 3 sizes: Small 12 x 12 x 20 in., Medium 12 x 11 x 38 in., Large 15 x 13 x 38 in.
- 5 boxes per case
- Quarterly report available upon request: Reports to include, but not limited to, materials recycled, amount processed, and materials end-of-life scenarios.
Specifications
82.20 L | |
12 x 11 x 38 in. | |
Not Autoclavable | |
Recycled Materials |
18.08 gal. | |
Medium | |
Cleanroom Socks | |
Recycling Box |
Provide Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction