Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Eisco™ Class A Volumetric Flasks with Individual Work Certificates, USP
SCI_ED

Class A Volumetric Flasks are USP-certified, and include interchangeable polypropylene stoppers and individual work certificates.

$12.50 - $79.00

Specifications

Closure Material Polypropylene
Material Borosilicate Glass 3.3
For Use With (Application) Preparing Solutions
Graduations Single, White Graduation Mark
Includes Individual Work Certificate
View More Specs

Products 10
Catalog Number Mfr. No. Capacity (Metric) Price Quantity & Availability  
Catalog Number Mfr. No. Capacity (Metric) Price Quantity & Availability  
S15083
SCI_ED
View Documents
Eisco™
CH1972A
5 mL
Each for $12.50
Add to Cart
 
S15084
SCI_ED
View Documents
Eisco™
CH1972B
10 mL
Each for $12.50
Add to Cart
 
S15085
SCI_ED
View Documents
Eisco™
CH1972C
25 mL
Each for $14.50
Add to Cart
 
S15086
SCI_ED
View Documents
Eisco™
CH1972D
50 mL
Each for $17.75
Add to Cart
 
S15087
SCI_ED
View Documents
Eisco™
CH1972E
100 mL
Each for $20.25
Add to Cart
 
S15088
SCI_ED
View Documents
Eisco™
CH1972F
200 mL
Each for $22.50
Add to Cart
 
S15089
SCI_ED
View Documents
Eisco™
CH1972G
250 mL
Each for $18.25
Add to Cart
 
S15090
SCI_ED
View Documents
Eisco™
CH1972H
500 mL
Each for $31.75
Add to Cart
 
S15091
SCI_ED
View Documents
Eisco™
CH1972I
1000 mL
Each for $42.25
Add to Cart
 
S15092
SCI_ED
View Documents
Eisco™
CH1972J
2000 mL
Each for $79.00
Add to Cart
 
Description

Description

  • Constructed of high quality, heavy duty Borosilicate 3.3 glass
  • Calibrated to contain, with a single graduation mark
  • Fitted with a chemical-resistant polypropylene stopper
  • Flask and stopper are microwavable and autoclavable
  • Low temperature gradient, capable of withstanding high temperatures & thermal shocks
  • Individual work certificates included in each box
  • Superior chemical durability
  • Class A tolerance
  • USP-certified
Specifications

Specifications

Polypropylene
Preparing Solutions
Individual Work Certificate
Uncoated
No
Beaded
A
Stopper
Borosilicate Glass 3.3
Single, White Graduation Mark
Volumetric Flask
Autoclavable
Yes
Non-sterile
Flat
SDS
Documents

Documents

Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit