Learn More
Fisherbrand™ Class 100 Vinyl Clean Room Chair - Medium Bench Height with Seat Tilt, Chrome Foot Ring, and Stationary Glides in Grade B Vinyl
SDPSCI_ED
Fisherbrand™ Class 100 Vinyl Clean Room Chairs are height adjustable for 36 in. work surfaces and feature ergonomic adjustments, clean room components, foot ring, and stationary glides. Antibacterial/Antifungal Grade B vinyl is easily cleaned making it ideal for clean room environments.
$427.00
Specifications
Adjustable Yes
Adjustable (Height) 21.5 to 29 in.
Back Style Standard
Base Material Polished Cast Aluminum
Certifications/Compliance Class 100/ISO Class 5
View More Specs
Products 5
Catalog Number Color Price Quantity & Availability  
Catalog Number Color Price Quantity & Availability  
S36986
SDPSCI_ED
View Documents
Black
Each for $427.00
Add to Cart
S36920
SDPSCI_ED
View Documents
Blue
Each for $427.00
Add to Cart
S36987
SDPSCI_ED
View Documents
Burgundy
Each for $427.00
Add to Cart
S36923
SDPSCI_ED
View Documents
Gray
Each for $427.00
Add to Cart
S36805
SDPSCI_ED
View Documents
Dark Blue
Each for $427.00
Add to Cart
Description
Description
  • Adjustable from 21.5-29 inches from the top of the seat to the floor
  • Features ergonomic adjustments including seat height
  • Locking back tilt
  • 8-degree forward and 3-degree backward locking seat tilt
  • 20-Inch Chrome adjustable foot ring with internal lock
  • Cast-aluminum base
  • Stationary, non-marring glides suitable for hard floors
  • Antibacterial/antifungal Grade B Vinyl
  • Variety of color options to suit any work space
  • Class 100/ISO 5 Clean Room
Specifications
Specifications
Yes
Standard
Class 100/ISO Class 5
Black-non-marring bell glides (suitable for both hard and carpeted floors)
300 lb.
Medium Bench Height
Yes
Yes
Cleanroom Chair
21.5 to 29 in.
Polished Cast Aluminum
20 in.
Round tube chrome foot ring with internal lock
136 kg
No
No
Yes
Vinyl