Learn More
Fisherbrand™ Class 10 Clean Room Vinyl Chair Chrome - Desk Height with Casters in Grade B Vinyl
SDPSCI_ED
Fisherbrand™ Class 10 Clean Room Chairs are height adjustable for 31 in. work surfaces and feature clean room components and self-braking casters. Antibacterial/Antifungal Grade B vinyl is easily cleaned making it ideal for clean room environments.
$335.00
Specifications
Adjustable Yes
Adjustable (Height) 18.5 to 23.75 in.
Back Style Standard
Base Material Polished Cast Aluminum
Certifications/Compliance Class 10/ISO Class 4
View More Specs
Products 5
Catalog Number Color Price Quantity & Availability  
Catalog Number Color Price Quantity & Availability  
S36591
SDPSCI_ED
View Documents
Blue
Each for $335.00
Add to Cart
S36934
SDPSCI_ED
View Documents
Black
Each for $335.00
Add to Cart
S36935
SDPSCI_ED
View Documents
Burgundy
Each for $335.00
Add to Cart
S36590
SDPSCI_ED
View Documents
Charcoal
Each for $335.00
Add to Cart
S36990
SDPSCI_ED
View Documents
Imperial Blue
Each for $335.00
Add to Cart
Description
Description
  • Adjustable from 18.5-23.75 inches from the top of the seat to the floor
  • Features manual back height and back depth adjustments
  • Cast-aluminum base
  • Chrome-hooded, dual-wheel, self-braking, non-marring casters suitable for hard floors
  • Antifungal/antibacterial Grade B vinyl
  • Variety of color options to suit any work space
  • Class 10/ISO 4 Clean Room
Specifications
Specifications
Yes
Standard
Class 10/ISO Class 4
300 lb.
Desk Height
Yes
No
Cleanroom Chair
18.5 to 23.75 in.
Polished Cast Aluminum
Chrome-hooded dual-wheel, self-braking, non-marring casters (suitable for both hard and carpeted floors)
136 kg
No
Yes
No
Vinyl