Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Cinnabarinic acid, Tocris Bioscience™
Click to view available options
Quantity:
10 mg
50 mg
Description
mGlu4 receptor agonist. Activates mGlu4 receptors in transiently transfected HEK293 cells; selective over all other mGlu subtypes (mGlu1, mGlu2 and mGlu5-8). Decreases cAMP levels in native cultured cerebellar granule cells. Also counteracts excitotoxic neuronal cell death induced by NMDA in mixed cultures of cortical cells; displays neuroprotective activity.
Specifications
Quantity | 50 mg |
Formula Weight | 300.22 |
Percent Purity | >98% |
Chemical Name or Material | Cinnabarinic acid |
Safety and Handling
Recommended Storage | Store at -20°C |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction