Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Chums™ Cotton Safety Chum Dual Breakaway Eyewear Retainers
Click to view available options
Breakaway with dual release clips; Black
Description
- Slider bead ensures a secure, reliable fit
- Available in a variety of colors
- Logo option available
- Made in the USA
Specifications
Color | Black |
Product Type | Dual Breakaway Eyewear Retainer |
For Use With (Equipment) | Designed for center-punched eyewear |
Includes | dual release clips |
Provide Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction