Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ChromaCare™ LC-MS Instrument Flush Solution, Thermo Scientific™
ChromaCare™ LC-MS Instrument Flush Solution is designed to prepare LC/MS instruments for start-up. By reducing background noise, this solution facilitates instrument installation and preventative maintenance routines.
Supplier: Fisher Chemical T111102500
Description
- Designed by Thermo Scientific to maximize cleaning while preserving instrument integrity in Vanquish™ and UltiMate™ UHPLC systems with QQQ MS
- Comprised of a unique blend of polar protic and aprotic solvents for removing a wide variety of contaminants
- Particularly suitable for new LC/MS and UHPLC/MS installations to rapidly stabilize baseline
- A blend of 25% acetonitrile, 25% methanol, 25%, water, and 25% 2-propanol (IPA)
- Offered in 1L borosilicate bottles and 2.5L amber glass bottles
Specifications
Liquid | |
25% Acetonitrile, 25% Methanol, 25% Water, 25% 2-Propanol (IPA) | |
Amber glass bottle | |
2.5 L |
75-05-8; 67-56-1; 67-63-0; 7732-18-5 | |
LC-MS | |
Colorless | |
Calcium (Ca): ≤ 20 ppb Sodium (Na): ≤ 50 ppb |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
RUO – Research Use Only