Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Chloroform (Ethanol as Preservative/Certified ACS), Fisher Chemical™
Click to view available options
Quantity:
500 mL
1 L
4 L
20 L
200 L
Packaging:
Aluminum Can
Glass Bottle
Safe-Cote™ Amber Glass
Steel Drum
Steel Pail
Specifications
Melting Point | -63°C |
Color | Colorless |
Decomposition Information | 290°C |
Density | 1.480g/cm³ |
Boiling Point | 61°C |
Quantity | 4 L |
Identification | Pass Test |
Packaging | Safe-Cote™ Amber Glass |
Physical Form | Liquid |
Residue after Evaporation | 0.001% max. |
Show More |
Safety and Handling
Hazard Statement | WARNING! |
Precautionary Statement | Emergency Overview Possible cancer hazard. May cause cancer based on animal data. Irritating to eyes and skin. May cause central nervous system effects. Aspiration hazard if swallowed - can enter lungs and cause damage. May be harmful if inhaled. May cause irritation of respiratory tract. Danger of serious damage to health by prolonged exposure. Use personal protective equipment. Use only under a chemical fume hood. Wash off immediately with plenty of water for at least 15 minutes. Obtain medical attention. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Obtain medical attention. NFPA Health:2 Flammability:1 Instability:0 |
DOTInformation | DOT Class 6.1, : Poison |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction