Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mopec Chlorine Free Body Bag with Black Envelope Zipper
Mopec Chlorine Free Body Bag with Black Envelope Zipper has curved black zipper and is suitable for cremation
Supplier: Mopec BE066
Description
- Features envelope style zipper with two pulls
- No handles
- Minimum 5 year shelf life
- Remains flexible without cracking to -70°F. (-57°C) for winter environments or morgue coolers
Specifications
Chlorine free | |
Zip Closure | |
PEVA | |
-70°F to +180°F | |
94 x 36in. |
300 lb. | |
White | |
1 | |
8 mil |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction