Learn More
HANNA INSTRUMENTS INC Free and Total Chlorine CAL Check™ Standards
Supplier: HANNA INSTRUMENTS INC HI9341411
The HI93414-11 Free and Total Chlorine CAL Check™ Standards provide a simple solution to calibrating and validating compatible benchtop and portable photometers. This high quality set of standards is manufactured in our state-of-the-art facility and comes supplied with a certificate of analysis. The certificate of analysis provides the lot number, reference values, and expiration date for traceability when certifying compatible free and total chlorine photometers.Supplied with certificate of analysisStandard value of 1.00 ± 0.03 mg/L chlorine @ 25°CReusable with long shelf life stability
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.