Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Ergodyne™ Chill-Its™ 6665 Evaporative Cooling Vest
Click to view available options
Color:
Gray
Lime
Size:
Medium
Large
X-Large
2X-Large
3X-Large
Description
- V-neck with zipper closure
- Quilted nylon outer layer
- Activated, polymer-embedded fabric inside
- Water-repellent liner and black cotton-poly elastic trim
- Lightweight, durable, and washable
- Soak in water 2-5 minutes to activate
- Remains hydrated for up to 4 hours
- Mesh side panels offer improved ventilation and stretch
Specifications
Material | Nylon |
Product Type | Vest |
Size | Medium |
Color | Lime |
For Use With (Application) | Construction, Maintenance, Landscaping, Material Handling |
Quantity | 6/Pk. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction