Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Innovating Science™ Chemistry - Investigating Your World ACS Kit
SCI_ED

Developed by the American Chemical Society through grants from the National Science Foundation and National Institutes of Health. Innovating Science™ Chemistry - Investigating Your World ACS Kit gets students to investigate chemical changes and how

Supplier:  Innovating Science™ IS2550

Catalog No. S42862


Add to Cart

Alert:

WARNING: This product can expose you to chemicals including Phenolphthalein/ Methanol/ Methyl Isobutyl Ketone and Phthalates, which are known to the State of California to cause cancer and reproductive harm.


Description

Description

  • This series of activities is comprised of four lessons that investigate chemical changes
  • Using the instruction manual obtained from the American Chemical Society’s website, and the materials included in the kit, students will take an imaginary trip around the globe to meet scientists and get a sense of the wide variety of ways that chemistry is used to benefit mankind
  • Activities include the production of a gas, chemistry and color change, formation of a precipitate, and temperature change of a chemical reactions
  • Contains enough materials for eight groups.
  • Kit Includes:
    • Citric Acid (100 g)
    • Sodium Carbonate (100 g)
    • Sodium Bicarbonate (250 g)
    • Calcium Chloride (250 g)
    • Universal Indicator (8 x 25 mL)
    • Dish Detergent (30 mL)
    • Bromothymol Blue (30 mL)
    • 3 oz. cups (16)
    • 2 oz. cups (32)
    • 9 Medicine Cups
    • 18 Scoops
    • Toothpicks
    • 8 Student Thermometers
    • 8 Vials
    • 8 Spot Plates
    • 33 Pipettes
    • 1 Test Tube w/Rubber Stopper
    • 1 Balloon
    • 2 Drinking Straw
    • 8 Universal Indicator Chart
    • 6 Self Inflating Reaction Bag
    • 1 Foot Warmer (2 pack)
    • 18 Spoons
Specifications

Specifications

Product SDSs
White
13 to 20
1 Kit
General Chemistry
7 to 14
Science Education
13 x 10 x 7 in.
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit