Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MilliporeSigma™ ChemiSCREEN™ Recombinant Human Metabotropic Glutamate Receptor
Used in membrane preparation for Radioligand binding Assays and GTPγS binding
Supplier: MilliporeSigma™ HTS146M
Description
- Host Cells: Chem-1
- Protein Target: mGlu2
- Target Sub-Family: Glutamate (Metabotropic)
Specifications
Membranes | |
200 U/Kit |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction