Learn More
SECURALL PRODUCTS CHEM STORAGE LOCK W/OUT EXPLOS
SDP
Supplier:  SECURALL PRODUCTS B1200
Encompass_Preferred
Chemical Storage Locker W/ out explosion Panel8'4"H x 7'W x 9'DIncludes 42" x 80" DoorSump Capacity: 206 gals.
Catalog No. NC1595078
Item Discontinued This item has been discontinued and is no longer available. Please reach out to your sales representative.