Learn More
LECO Corporation Ceramic Boats Unglazed Pack of 250
Supplier: LECO Corporation 528203250
Unglazed, Round Bottom Shape. For macrosamples/horizontal-loading furnaces. For use with 632 Series, 628 Series Module, TruSpec Series Module, TruMac Series. White, Ceramic, Round Bottom, Max Temp 1800 deg., 65 x 21.5 x 11.5. 250/pk.