Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CEP Oil Gator Absorbent
An absorbent powder for any liquid; ideal for hydrocarbons
Supplier: CEP GATOR30
Description
- Cost-effective and environmentally friendly
- 14 kg bag
Specifications
Oil Gator Absorbent | |
Any liquid and is ideal for hydrocarbons | |
13.6 kg |
Granular | |
30 lb. |
Provide Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction