Learn More
Sarstedt Inc Screw cap tube, 15 ml, (LxØ): 120 x 17 mm, PS, with print, 500/case
Supplier: Sarstedt Inc 62.553.205
15mL Screw cap tubes come with a red cap assembles. They are sterile, non-pyrogenic/endotoxin-free, and non-cytotoxic. These tubes feature 2-color printed graduations and writing space.