Learn More
Cenmed™ Enterprises URITRAC™ Testing Kit
SDP
Semi-quantitative culture test for the detection of urinary tract infections
Supplier:  Cenmed™ 201201U
Catalog No. 21-520-104
Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.
Description
Description
  • Features a dip paddle method, thick media, and nail heads on the paddle
  • No waiting time between collection of samples and inoculation of media
  • Screens out negatives easily, with faster negative result reporting
  • Saves time and money by eliminating the work of labeling and streaking out 2 media plates
Specifications
Specifications
For the detection of urinary tract infections
100 Case